CAS RN 5486-55-5
CAS RN 5486-55-5
IUPAC Name:
2,6-dimethoxynaphthalene
Synonyms:
2,6-dimethoxynaphthalene
Molecular Weight (g/mol):
188.23
Molecular Formula:
C12H12O2
InChi Key:
AHKDVDYNDXGFPP-UHFFFAOYSA-N
SMILES:
COC1=CC2=CC=C(OC)C=C2C=C1
1
–
1
of
1
results
1
2,6-Dimethoxynaphthalene, 99%
CAS: 5486-55-5 Molecular Formula: C12H12O2 Molecular Weight (g/mol): 188.23 MDL Number: MFCD00004063 InChI Key: AHKDVDYNDXGFPP-UHFFFAOYSA-N Synonym: naphthalene, 2,6-dimethoxy,2,6-dimethoxy-naphthalene,2,6-dimethoxy naphthalene,pubchem22734,naphthalene,6-dimethoxy,acmc-1ap1a,2,6-dimethoxynaphthalene PubChem CID: 79627 IUPAC Name: 2,6-dimethoxynaphthalene SMILES: COC1=CC2=CC=C(OC)C=C2C=C1