accessibility menu, dialog, popup

UserName

CAS RN 548472-68-0

CH3OOCH3CH3NNONHNOClCl

CAS RN 548472-68-0

IUPAC Name: 4-[4,5-bis(4-chlorophenyl)-2-[4-methoxy-2-(propan-2-yloxy)phenyl]-4,5-dihydro-1H-imidazole-1-carbonyl]piperazin-2-one
Synonyms: nutlin
Molecular Weight (g/mol): 581.49
Molecular Formula: C30H30Cl2N4O4
InChi Key: BDUHCSBCVGXTJM-UHFFFAOYNA-N
SMILES: COC1=CC=C(C2=NC(C(N2C(=O)N2CCNC(=O)C2)C2=CC=C(Cl)C=C2)C2=CC=C(Cl)C=C2)C(OC(C)C)=C1
Color 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)