accessibility menu, dialog, popup

UserName

CAS RN 5464-11-9

HIH3CSNHN

CAS RN 5464-11-9

IUPAC Name: hydrogen 2-(methylsulfanyl)-4,5-dihydro-1H-imidazole iodide
Synonyms: hydrogen 2-(methylsulfanyl)-4,5-dihydro-1H-imidazole iodide
Molecular Weight (g/mol): 244.09
Molecular Formula: C4H9IN2S
InChi Key: PZZRSEUDGCFXIH-UHFFFAOYSA-N
SMILES: [H+].[I-].CSC1=NCCN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results