accessibility menu, dialog, popup

UserName

CAS RN 5461-08-5

H3CCH3OOOO

CAS RN 5461-08-5

IUPAC Name: (2H-1,3-benzodioxol-5-yl)methyl 2-methylpropanoate
Synonyms: 2H-1,3-benzodioxol-5-ylmethyl 2-methylpropanoate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: RQULTIASPCVEFO-UHFFFAOYSA-N
SMILES: CC(C)C(=O)OCC1=CC=C2OCOC2=C1
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results