accessibility menu, dialog, popup

UserName

CAS RN 5445-51-2

IUPAC Name: cyclobutane-1,1-dicarboxylic acid
Synonyms: H2cbdca
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: CCQPAEQGAVNNIA-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCC1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results