CAS RN 540737-29-9
IUPAC Name:
2-hydroxypropane-1,2,3-tricarboxylic acid; 3-{4-methyl-3-[methyl({7H-pyrrolo[2,3-d]pyrimidin-4-yl})amino]piperidin-1-yl}-3-oxopropanenitrile
Synonyms:
citric acid; tofacitinib
Molecular Weight (g/mol):
504.5
Molecular Formula:
C22H28N6O8
InChi Key:
SYIKUFDOYJFGBQ-UHFFFAOYNA-N
SMILES:
OC(=O)CC(O)(CC(O)=O)C(O)=O.CC1CCN(CC1N(C)C1=C2C=CNC2=NC=N1)C(=O)CC#N
1
–
3
of
3
results
1
Thermo Scientific Chemicals Tofacitinib citrate
CAS: 540737-29-9 Molecular Formula: C16H20N6O·C6H8O7
2
Tofacitinib Citrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
R&D Systems™ CP 690550 citrate
CAS: 540737-29-9 Molecular Formula: C22H28N6O8