accessibility menu, dialog, popup

UserName

CH3OH3COH3C

CAS RN 534-15-6

IUPAC Name: 1,1-dimethoxyethane
Synonyms: 1,1-dimethoxyethane
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: SPEUIVXLLWOEMJ-UHFFFAOYSA-N
SMILES: COC(C)OC
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results