accessibility menu, dialog, popup

UserName

CAS RN 53312-77-9

ClH2NN

CAS RN 53312-77-9

IUPAC Name: 2-amino-3-chlorobenzonitrile
Synonyms: 2-amino-3-chlorobenzonitrile
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: FAHVRFGAGJMXHW-UHFFFAOYSA-N
SMILES: NC1=C(C=CC=C1Cl)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results