accessibility menu, dialog, popup

UserName

CAS RN 530-55-2

CH3OOOCH3O

CAS RN 530-55-2

IUPAC Name: 2,6-dimethoxycyclohexa-2,5-diene-1,4-dione
Synonyms: 2,6-dimethoxy-1,4-benzoquinone 2,6-dimethoxy-P-benzoquinone
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: OLBNOBQOQZRLMP-UHFFFAOYSA-N
SMILES: COC1=CC(=O)C=C(OC)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)