accessibility menu, dialog, popup

UserName

CAS RN 52988-34-8

IUPAC Name: 4'-methoxy-[1,1'-biphenyl]-4-carbaldehyde
Synonyms: 4'-methoxy-[1,1'-biphenyl]-4-carbaldehyde
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: JTTIGLYPLMYHAT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results