accessibility menu, dialog, popup

UserName

CAS RN 52829-98-8

H3COH

CAS RN 52829-98-8

IUPAC Name: 1-cyclopentylethan-1-ol
Synonyms: 1-cyclopentylethanol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: WPCMSUSLCWXTKB-UHFFFAOYNA-N
SMILES: CC(O)C1CCCC1
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)