accessibility menu, dialog, popup

UserName

CAS RN 521937-07-5

ONHOHO

CAS RN 521937-07-5

IUPAC Name: 4-hydroxy-8-oxa-11-azatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraen-10-one
Synonyms: 4-hydroxy-8-oxa-11-azatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraen-10-one
Molecular Weight (g/mol): 217.22
Molecular Formula: C12H11NO3
InChi Key: AACFPJSJOWQNBN-UHFFFAOYSA-N
SMILES: OC1=CC=C2OC3=C(CCCNC3=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results