accessibility menu, dialog, popup

UserName

CAS RN 51953-17-4

ONN

CAS RN 51953-17-4

IUPAC Name: 4,5-dihydropyrimidin-4-one
Synonyms: pyrimidone
Molecular Weight (g/mol): 96.09
Molecular Formula: C4H4N2O
InChi Key: TZYQTWHRLVDYPL-UHFFFAOYSA-N
SMILES: O=C1CC=NC=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)