accessibility menu, dialog, popup

UserName

CAS RN 519-05-1

H3COOHOOOCH3

CAS RN 519-05-1

IUPAC Name: 6-formyl-2,3-dimethoxybenzoic acid
Synonyms: 6-formyl-2,3-dimethoxybenzoic acid
Molecular Weight (g/mol): 210.19
Molecular Formula: C10H10O5
InChi Key: HVXXOIGTXJOVON-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C(C(O)=O)=C1OC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results