accessibility menu, dialog, popup

UserName

CAS RN 51451-44-6

IUPAC Name: 2-(pyridin-3-yl)ethanethioamide
Synonyms: 2-(pyridin-3-yl)ethanethioamide
Molecular Weight (g/mol): 152.22
Molecular Formula: C7H8N2S
InChi Key: VSBVYPLNYOJQEV-UHFFFAOYSA-N
SMILES: NC(=S)CC1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results