accessibility menu, dialog, popup

UserName

CAS RN 5144-10-5

CH3CH3CH3NH3CH3C

CAS RN 5144-10-5

IUPAC Name: pentamethylbenzonitrile
Synonyms: pentamethylbenzonitrile
Molecular Weight (g/mol): 173.26
Molecular Formula: C12H15N
InChi Key: NCDPYDGGUAKABA-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)=C(C#N)C(C)=C1C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results