accessibility menu, dialog, popup

UserName

H3COOOO

CAS RN 51264-68-7

IUPAC Name: ethyl 2-(3-formylphenoxy)acetate
Synonyms: ethyl 2-(3-formylphenoxy)acetate
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: HMBNSDWJNMBKAE-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=CC=CC(C=O)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results