accessibility menu, dialog, popup

UserName

CAS RN 51037-30-0

CH3NOHONO

CAS RN 51037-30-0

IUPAC Name: 2-carboxy-6-methylpyrazin-1-ium-1-olate
Synonyms: acipimox
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: DNRXJHATQULEHC-UHFFFAOYSA-N
SMILES: CC1=CN=CC(C(O)=O)=[N+]1[O-]
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)