accessibility menu, dialog, popup

UserName

FNNHNN

CAS RN 50907-20-5

IUPAC Name: 5-(3-fluorophenyl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(3-fluorophenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 164.14
Molecular Formula: C7H5FN4
InChi Key: SEYSTLGMSOLFBD-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results