accessibility menu, dialog, popup

UserName

CAS RN 502496-23-3

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]hydrazine hydrochloride
Synonyms: [3,5-bis(trifluoromethyl)phenyl]hydrazine hydrochloride
Molecular Weight (g/mol): 280.6
Molecular Formula: C8H7ClF6N2
InChi Key: OPIOKSFIERNABH-UHFFFAOYSA-N
SMILES: Cl.NNC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results