accessibility menu, dialog, popup

UserName

CAS RN 497934-76-6

N(R)(R)NN(R)(R)OOPPO(R)(R)(S)(S)NNOOOOOOOHHOONH2NH2OHHONaNaH2OH2OH2O

CAS RN 497934-76-6

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results