accessibility menu, dialog, popup

UserName

CAS RN 4938-52-7

H3COHCH2

CAS RN 4938-52-7

IUPAC Name: hept-1-en-3-ol
Synonyms: hept-1-en-3-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: PZKFYTOLVRCMOA-UHFFFAOYNA-N
SMILES: CCCCC(O)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)