CAS RN 487-58-1
CAS RN 487-58-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hypaphorine, TRC
CAS: 487-58-1 Molecular Formula: C14H18N2O2 Molecular Weight (g/mol): 246.31 Synonym: 1H-Indole-3-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (αS)- (9CI, ACI),Hypaphorine (6CI),(1-Carboxy-2-indol-3-ylethyl)trimethylammonium hydroxide, inner salt (7CI),1H-Indole-3-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (S)- (ZCI),Ammonium, (1-carboxy-2-indol-3-ylethyl)trimethyl-, hydroxide, inner salt (8CI),(+)-Hypaphorine,L-Hypaphorine,Glyyunnanenine,Lenticin,Lenticine,Tryptophan betaine,Tryptophan, trimethylbetaine IUPAC Name: (2S)-3-(1H-indol-3-yl)-2-(trimethylazaniumyl)propanoate SMILES: C[N+](C)(C)[C@@H](Cc1c[nH]c2ccccc12)C(=O)[O-]