CAS RN 482-36-0
CAS RN 482-36-0
IUPAC Name:
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
Synonyms:
hyperoside quercetin-3-O-galactoside quercetin-3-galactoside quercetin 3-galactoside hyperosid quercetin galactoside hyperozide hyperasid hyperin (8CI)
Molecular Weight (g/mol):
464.38
Molecular Formula:
C21H20O12
InChi Key:
OVSQVDMCBVZWGM-DTGCRPNFSA-N
SMILES:
OC[C@H]1O[C@@H](OC2=C(OC3=CC(O)=CC(O)=C3C2=O)C2=CC=C(O)C(O)=C2)[C@H](O)[C@@H](O)[C@H]1O
1
–
3
of
3
results
1
Hyperoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Hyperoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Hyperoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.