accessibility menu, dialog, popup

UserName

CAS RN 4812-20-8

CH3H3COHO

CAS RN 4812-20-8

IUPAC Name: 2-(propan-2-yloxy)phenol
Synonyms: 2-isopropoxyphenol caswell no. 508A epa pesticide chemical code 205400
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: ZNCUUYCDKVNVJH-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)