accessibility menu, dialog, popup

UserName

CAS RN 480-91-1

OHN

CAS RN 480-91-1

IUPAC Name: 2,3-dihydro-1H-isoindol-1-one
Synonyms: 2,3-dihydro-1H-isoindol-1-one
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: PXZQEOJJUGGUIB-UHFFFAOYSA-N
SMILES: O=C1NCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results