CAS RN 478336-92-4
CAS RN 478336-92-4
IUPAC Name:
methyl 2-[3-(4-oxocyclohexa-2,5-dien-1-ylidene)-1,2-oxazolidin-5-yl]acetate
Synonyms:
methyl 2-[3-(4-oxocyclohexa-2,5-dien-1-ylidene)-1,2-oxazolidin-5-yl]acetate
Molecular Weight (g/mol):
235.24
Molecular Formula:
C12H13NO4
InChi Key:
CLVQGFPQFNASJH-UHFFFAOYNA-N
SMILES:
COC(=O)CC1CC(NO1)=C1C=CC(=O)C=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals MIF Antagonist, ISO-1
CAS: 478336-92-4 Molecular Formula: C12H13NO4,C12H13NO4 Synonym: (S,R)-3-(4-Hydroxyphenyl)-4,5-dihydro-5-isoxazole acetic acid, methyl ester; Macrophage Migration Inhibitory Factor Antagonist
2
Tocris Bioscience™ ISO 1
CAS: 478336-92-4 Molecular Formula: C12H13NO4