CAS RN 4569-88-4
CAS RN 4569-88-4
IUPAC Name:
3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Synonyms:
3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxonaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular Weight (g/mol):
506.01
Molecular Formula:
C30H24ClN5O
InChi Key:
SCMDRBZEIUMBBQ-UHFFFAOYSA-N
SMILES:
[Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12
1
–
1
of
1
results
1
Indoine Blue, Thermo Scientific™
CAS: 4569-88-4 Molecular Formula: C30H24ClN5O Molecular Weight (g/mol): 506.01 MDL Number: MFCD00011924 InChI Key: SCMDRBZEIUMBBQ-UHFFFAOYSA-N PubChem CID: 9575841 SMILES: [Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12