accessibility menu, dialog, popup

UserName

CAS RN 45000-93-9

H3COHH3COHH3COHClWCl

CAS RN 45000-93-9

IUPAC Name: dichlorotungsten; tris(ethanol)
Synonyms: dichlorotungsten; tris(ethyl alcohol)
Molecular Weight (g/mol): 392.95
Molecular Formula: C6H18Cl2O3W
InChi Key: AQDCOTYKQNGVLX-UHFFFAOYSA-L
SMILES: CCO.CCO.CCO.Cl[W]Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results