CAS RN 4449-51-8
CAS RN 4449-51-8
IUPAC Name:
(3S,3'R,3'aS,6'S,6aS,6bS,7'aR,9R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3'H-spiro[cyclohexa[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol
Synonyms:
cyclopamine 11-deoxyjervine 11-deoxojervine 11-deoxo- (7CI) jervine jervine, 11-deoxo- (7CI)
Molecular Weight (g/mol):
411.63
Molecular Formula:
C27H41NO2
InChi Key:
QASFUMOKHFSJGL-LAFRSMQTSA-N
SMILES:
C[C@@H]1[C@@H]2NC[C@@H](C)C[C@H]2O[C@]11CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC2=C1C
1
–
3
of
3
results
1
Cyclopamine, 99%
CAS: 4449-51-8 Molecular Formula: C27H41NO2 Molecular Weight (g/mol): 411.63 MDL Number: MFCD01735266 InChI Key: QASFUMOKHFSJGL-LAFRSMQTSA-N Synonym: cyclopamine,3b,23b-17,23-epoxy-11-deoxoveratraman-3-ol PubChem CID: 122130428 SMILES: C[C@@H]1[C@@H]2NC[C@@H](C)C[C@H]2O[C@]11CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC2=C1C
3