CAS RN 444728-11-4
CAS RN 444728-11-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
rac-2-(2-Chlorophenyl)-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
rac-2-(2-Chlorophenyl)-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
rac-2-(2-Chlorophenyl)-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.