accessibility menu, dialog, popup

UserName

CAS RN 4432-31-9

OHSOONO

CAS RN 4432-31-9

IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid
Synonyms: mes (buffer) MES mes (buffering agent)
Molecular Weight (g/mol): 195.23
Molecular Formula: C6H13NO4S
InChi Key: SXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES: OS(=O)(=O)CCN1CCOCC1
Molecular Weight (g/mol) 
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)