CAS RN 4405-13-4
CAS RN 4405-13-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Hexahydro-[1,4]dioxino[2,3-b][1,4]dioxine-2,3,6,7-tetrol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Hexahydro-[1,4]dioxino[2,3-b][1,4]dioxine-2,3,6,7-tetrol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Hexahydro-[1,4]dioxino[2,3-b][1,4]dioxine-2,3,6,7-tetrol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.