accessibility menu, dialog, popup

UserName

CAS RN 43163-94-6

H3CCH3FNH2OHO

CAS RN 43163-94-6

IUPAC Name: (2S)-2-azaniumyl-3-fluoro-3-methylbutanoate
Synonyms: (2S)-2-ammonio-3-fluoro-3-methylbutanoate
Molecular Weight (g/mol): 135.14
Molecular Formula: C5H10FNO2
InChi Key: ZFUKCHCGMBNYHH-VKHMYHEASA-N
SMILES: CC(C)(F)[C@@H]([NH3+])C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)