accessibility menu, dialog, popup

UserName

CAS RN 43100-38-5

CH3H3CCH3OHNNH2

CAS RN 43100-38-5

IUPAC Name: 4-tert-butylbenzohydrazide
Synonyms: 4-tert-butylbenzohydrazide
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: XYUFQWDLRLHUPB-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)NN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results