CAS RN 4287-30-3
CAS RN 4287-30-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,5-Dimethylphenoxymethyloxirane, TRC
CAS: 4287-30-3 Molecular Formula: C11H14O2 Molecular Weight (g/mol): 178.23 Synonym: [(3,5-Dimethylphenoxy)methyl]oxirane,1,2-Epoxy-3-(3,5-xylyloxy)propane,1-(3,5-Dimethylphenoxy)-2,3-epoxypropane,Glycidyl 3,5-Dimethylphenyl Ether SMILES: OCC(C)(C)NC(C)C(=O)c1ccc(F)cc1