accessibility menu, dialog, popup

UserName

CAS RN 423165-33-7

CH3ONOOBr

CAS RN 423165-33-7

IUPAC Name: 1-bromo-2-ethoxy-4-nitrobenzene
Synonyms: 1-bromo-2-ethoxy-4-nitrobenzene
Molecular Weight (g/mol): 246.06
Molecular Formula: C8H8BrNO3
InChi Key: ZALXYXWTUXLPBJ-UHFFFAOYSA-N
SMILES: CCOC1=C(Br)C=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results