accessibility menu, dialog, popup

UserName

CAS RN 4224-69-5

H3COOCH2Br

CAS RN 4224-69-5

IUPAC Name: methyl 2-(bromomethyl)prop-2-enoate
Synonyms: methyl 2-(bromomethyl)prop-2-enoate
Molecular Weight (g/mol): 179.01
Molecular Formula: C5H7BrO2
InChi Key: CFTUQSLVERGMHL-UHFFFAOYSA-N
SMILES: COC(=O)C(=C)CBr
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results