accessibility menu, dialog, popup

UserName

CAS RN 42228-92-2

H2N(S)(S)NOClOHO

CAS RN 42228-92-2

IUPAC Name: (2S)-2-amino-2-[(5S)-3-chloro-4,5-dihydro-1,2-oxazol-5-yl]acetic acid
Synonyms: acivicin 阿西维辛 C5H7ClN2O3 acivicine acivicino acivicinum antibiotic AT 125
Molecular Weight (g/mol): 178.57
Molecular Formula: C5H7ClN2O3
InChi Key: QAWIHIJWNYOLBE-OKKQSCSOSA-N
SMILES: N[C@@H]([C@@H]1CC(Cl)=NO1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results