CAS RN 4206-67-1
IUPAC Name:
(iodomethyl)trimethylsilane
Synonyms:
(iodomethyl)trimethylsilane
Molecular Weight (g/mol):
214.12
Molecular Formula:
C4H11ISi
InChi Key:
VZNYXGQMDSRJAL-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)CI
1
–
1
of
1
results
1
(Iodomethyl)trimethylsilane, 99%
CAS: 4206-67-1 Molecular Formula: C4H11ISi Molecular Weight (g/mol): 214.121 MDL Number: MFCD00001077 InChI Key: VZNYXGQMDSRJAL-UHFFFAOYSA-N Synonym: iodomethyl trimethylsilane,iodomethyl-trimethylsilane,iodomethyl trimethyl silane,trimethylsilyl methyl iodide,sila-neopentyliodide,trimethylsilylmethyl iodide,acmc-209jn7,trimethyl iodomethyl silane,iodanylmethyl trimethyl silane PubChem CID: 77877 IUPAC Name: iodomethyl(trimethyl)silane SMILES: C[Si](C)(C)CI