accessibility menu, dialog, popup

UserName

H3CHNOH

CAS RN 42055-15-2

IUPAC Name: 3-(methylamino)propan-1-ol
Synonyms: 3-(methylamino)propan-1-ol
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: KRGXWTOLFOPIKV-UHFFFAOYSA-N
SMILES: CNCCCO
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)