CAS RN 4137-10-4
CAS RN 4137-10-4
IUPAC Name:
dimethylamine; dimethylcarbamic acid
Synonyms:
carbamic acid, dimethyl-; dimethylamine
Molecular Weight (g/mol):
134.18
Molecular Formula:
C5H14N2O2
InChi Key:
JIYXHCMRGZVYMA-UHFFFAOYSA-N
SMILES:
CNC.CN(C)C(O)=O
1
–
1
of
1
results
1
Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1
CAS: 4137-10-4 Molecular Formula: C5H14N2O2 Molecular Weight (g/mol): 134.18 MDL Number: MFCD00029888 InChI Key: JIYXHCMRGZVYMA-UHFFFAOYSA-N Synonym: DIMCARB IUPAC Name: dimethylamine; dimethylcarbamic acid SMILES: CNC.CN(C)C(O)=O