accessibility menu, dialog, popup

UserName

CAS RN 41264-06-6

CH3OOCH3OOH3C

CAS RN 41264-06-6

IUPAC Name: methyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate
Synonyms: methyl 2,4-dimethyl-6-oxopyran-3-carboxylate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: VLYBFLKIZIACQH-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)OC(=O)C=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results