accessibility menu, dialog, popup

UserName

CAS RN 409351-28-6

CH3NSONHOCH3H2NCH3

CAS RN 409351-28-6

IUPAC Name: 3-amino-N-[(4-methoxyphenyl)methyl]-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide
Synonyms: 3-amino-N-[(4-methoxyphenyl)methyl]-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide
Molecular Weight (g/mol): 341.43
Molecular Formula: C18H19N3O2S
InChi Key: MDNWGCQSCGNTKH-UHFFFAOYSA-N
SMILES: COC1=CC=C(CNC(=O)C2=C(N)C3=C(C)C=C(C)N=C3S2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results