accessibility menu, dialog, popup

UserName

CAS RN 4093-31-6

CH3OOClNHCH3OOCH3

CAS RN 4093-31-6

IUPAC Name: methyl 5-chloro-4-acetamido-2-methoxybenzoate
Synonyms: methyl 5-chloro-4-acetamido-2-methoxybenzoate
Molecular Weight (g/mol): 257.67
Molecular Formula: C11H12ClNO4
InChi Key: OUEXNQRVYGYGIK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=C(NC(C)=O)C=C1OC
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results