accessibility menu, dialog, popup

UserName

CAS RN 402713-81-9

H3COSOONHOH3CClClNNHHCl

CAS RN 402713-81-9

IUPAC Name: N-(3,5-dichloro-2-methoxyphenyl)-4-methoxy-3-(piperazin-1-yl)benzene-1-sulfonamide hydrochloride
Synonyms: N-(3,5-dichloro-2-methoxyphenyl)-4-methoxy-3-(piperazin-1-yl)benzenesulfonamide hydrochloride
Molecular Weight (g/mol): 482.8
Molecular Formula: C18H22Cl3N3O4S
InChi Key: RNKCEBCFUSXSQE-UHFFFAOYSA-N
SMILES: Cl.COC1=C(C=C(C=C1)S(=O)(=O)NC1=CC(Cl)=CC(Cl)=C1OC)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results