accessibility menu, dialog, popup

UserName

H2CFFF

CAS RN 402-50-6

IUPAC Name: 1-ethenyl-4-(trifluoromethyl)benzene
Synonyms: 1-ethenyl-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 172.15
Molecular Formula: C9H7F3
InChi Key: CEWDRCQPGANDRS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)