CAS RN 39574-16-8
CAS RN 39574-16-8
IUPAC Name:
ethyl 2-isothiocyanatopropanoate
Synonyms:
ethyl 2-isothiocyanatopropanoate
Molecular Weight (g/mol):
159.2
Molecular Formula:
C6H9NO2S
InChi Key:
ALJGYASQFZQQJX-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(C)N=C=S
1
–
1
of
1
results
1
Ethyl 2-isothiocyanatopropionate, 97%
CAS: 39574-16-8 Molecular Formula: C6H9NO2S Molecular Weight (g/mol): 159.20 MDL Number: MFCD00070636 InChI Key: ALJGYASQFZQQJX-UHFFFAOYNA-N Synonym: ethyl 2-isothiocyanatopropionate,ethyl n-thioxomethylene alaninate,propanoic acid, 2-isothiocyanato-, ethyl ester,acmc-20al0z,acmc-20lue7,propanoic acid, 2-isothiocyanato-, ethyl ester, s,ethyl l-2-isothiocyanatopropanoate,alanine, n-carbonothioyl-, ethyl ester PubChem CID: 142361 IUPAC Name: ethyl 2-isothiocyanatopropanoate SMILES: CCOC(=O)C(C)N=C=S