accessibility menu, dialog, popup

UserName

CAS RN 395-01-7

FFO

CAS RN 395-01-7

IUPAC Name: 2,2-difluoro-1-phenylethan-1-one
Synonyms: 2,2-difluoro-1-phenylethanone
Molecular Weight (g/mol): 156.13
Molecular Formula: C8H6F2O
InChi Key: OLYKCPDTXVZOQF-UHFFFAOYSA-N
SMILES: FC(F)C(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results