accessibility menu, dialog, popup

UserName

CAS RN 3946-29-0

ClOCl

CAS RN 3946-29-0

IUPAC Name: 3-chloro-1-(4-chlorophenyl)propan-1-one
Synonyms: 3-chloro-1-(4-chlorophenyl)propan-1-one
Molecular Weight (g/mol): 203.06
Molecular Formula: C9H8Cl2O
InChi Key: AYFJBHFMQODYBC-UHFFFAOYSA-N
SMILES: ClCCC(=O)C1=CC=C(Cl)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results